Anti-ALDH16A1 Mouse Polyclonal Antibody

H00126133-B01P
10510-670EA 316.92 USD
10510-670
Anti-ALDH16A1 Mouse Polyclonal Antibody
Type: Primary
Antigen: ALDH16A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR