Anti-OSCAR Mouse Monoclonal Antibody [clone: 4C2]

H00126014-M06
10566-402EA 357.81 USD
10566-402
Anti-OSCAR Mouse Monoclonal Antibody [clone: 4C2]
Type: Primary
Antigen: OSCAR
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR