Anti-CALR3 Mouse Polyclonal Antibody

Supplier: Abnova
H00125972-B01P
10552-370EA 335 USD
10552-370
Anti-CALR3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CALR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR