Anti-COX6B2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00125965-D01P
10582-768EA 376 USD
10582-768
Anti-COX6B2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX6B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR