Anti-CLDND2 Mouse Polyclonal Antibody

H00125875-B01P
10553-604EA 316.92 USD
10553-604
Anti-CLDND2 Mouse Polyclonal Antibody
Type: Primary
Antigen: CLDND2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR