Anti-TBC1D16 Mouse Polyclonal Antibody

H00125058-B01P
10580-092EA 316.92 USD
10580-092
Anti-TBC1D16 Mouse Polyclonal Antibody
Type: Primary
Antigen: TBC1D16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR