Anti-SNX20 Mouse Monoclonal Antibody [clone: 4E9]

Supplier: Abnova
H00124460-M05
10601-704EA 335 USD
10601-704
Anti-SNX20 Mouse Monoclonal Antibody [clone: 4E9]
Antibodies
Type: Primary
Antigen: SNX20
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR