Anti-USP45 Mouse Monoclonal Antibody [clone: 1H2]

Supplier: Abnova
H00085015-M01
10605-500EA 335 USD
10605-500
Anti-USP45 Mouse Monoclonal Antibody [clone: 1H2]
Antibodies
Type: Primary
Antigen: USP45
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR