Anti-RERG Mouse Monoclonal Antibody [clone: 1C5]

Supplier: Abnova
H00085004-M03
10599-522EA 335 USD
10599-522
Anti-RERG Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
Type: Primary
Antigen: RERG
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR