Anti-PPP1R16A Mouse Polyclonal Antibody

Supplier: Abnova
H00084988-B02
10568-244EA 335 USD
10568-244
Anti-PPP1R16A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP1R16A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR