Anti-FBXL20 Mouse Monoclonal Antibody [clone: 2G1]

Supplier: Abnova
H00084961-M03
10593-328EA 335 USD
10593-328
Anti-FBXL20 Mouse Monoclonal Antibody [clone: 2G1]
Antibodies
Type: Primary
Antigen: FBXL20 (F-box and leucine-rich repeat protein 20)
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR