Anti-RELT Mouse Monoclonal Antibody [clone: 3F8]

Supplier: Abnova
H00084957-M01
10604-056EA 335 USD
10604-056
Anti-RELT Mouse Monoclonal Antibody [clone: 3F8]
Antibodies
Type: Primary
Antigen: RELT
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR