Anti-TNS4 Mouse Polyclonal Antibody

H00084951-B01P
10572-720EA 316.92 USD
10572-720
Anti-TNS4 Mouse Polyclonal Antibody
Type: Primary
Antigen: TNS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR