Anti-ZFYVE19 Mouse Polyclonal Antibody

Supplier: Abnova
H00084936-B01
10574-230EA 335 USD
10574-230
Anti-ZFYVE19 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZFYVE19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR