Anti-ARHGEF39 Mouse Polyclonal Antibody

H00084904-B01P
10552-190EA 316.92 USD
10552-190
Anti-ARHGEF39 Mouse Polyclonal Antibody
Type: Primary
Antigen: ARHGEF39 (Rho guanine nucleotide exchange factor (GEF) 39)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR