Anti-NFATC2IP Mouse Monoclonal Antibody [clone: 2D7-A10]

Supplier: Abnova
H00084901-M01
10586-208EA 335 USD
10586-208
Anti-NFATC2IP Mouse Monoclonal Antibody [clone: 2D7-A10]
Antibodies
Type: Primary
Antigen: NFATC2IP (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein)
Clonality: Monoclonal
Clone: 2D7-A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR