Anti-PLXDC2 Mouse Monoclonal Antibody [clone: 4G10]

Supplier: Abnova
H00084898-M01
10597-354EA 335 USD
10597-354
Anti-PLXDC2 Mouse Monoclonal Antibody [clone: 4G10]
Antibodies
Type: Primary
Antigen: PLXDC2
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR