Anti-AIFM2 Mouse Monoclonal Antibody [clone: 2C6]

H00084883-M13
10551-300EA 357.81 USD
10551-300
Anti-AIFM2 Mouse Monoclonal Antibody [clone: 2C6]
Type: Primary
Antigen: AIFM2
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR