Anti-AIFM2 Mouse Polyclonal Antibody

H00084883-B01P
10551-302EA 316.92 USD
10551-302
Anti-AIFM2 Mouse Polyclonal Antibody
Type: Primary
Antigen: AIFM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR