Anti-PLCD4 Mouse Monoclonal Antibody [clone: 4D4]

Supplier: Abnova
H00084812-M01
10597-232EA 335 USD
10597-232
Anti-PLCD4 Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
Type: Primary
Antigen: PLCD4
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR