Anti-FUT10 Mouse Polyclonal Antibody

Supplier: Abnova
H00084750-B01
10557-130EA 335 USD
10557-130
Anti-FUT10 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FUT10 (fucosyltransferase 10 (alpha (1,3) fucosyltransferase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR