Anti-CNDP1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00084735-D01P
10582-466EA 376 USD
10582-466
Anti-CNDP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNDP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR