Anti-ACCS Mouse Monoclonal Antibody [clone: 1D2]

Supplier: Abnova
H00084680-M02
10596-804EA 335 USD
10596-804
Anti-ACCS Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
Type: Primary
Antigen: ACCS
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR