Anti-TXLNG2P Mouse Monoclonal Antibody [clone: 2F5]

Supplier: Abnova
H00084663-M01
10583-336EA 335 USD
10583-336
Anti-TXLNG2P Mouse Monoclonal Antibody [clone: 2F5]
Antibodies
Type: Primary
Antigen: TXLNG2P
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR