Anti-NTNG2 Mouse Monoclonal Antibody [clone: 4F11]

Supplier: Abnova
H00084628-M01
10595-476EA 335 USD
10595-476
Anti-NTNG2 Mouse Monoclonal Antibody [clone: 4F11]
Antibodies
Type: Primary
Antigen: NTNG2
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR