Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 3A12]

Supplier: Abnova
H00084623-M02
10591-098EA 335 USD
10591-098
Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 3A12]
Antibodies
Type: Primary
Antigen: KIRREL3
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR