Anti-CAMK1D Mouse Monoclonal Antibody [clone: 3H8]

Supplier: Abnova
H00057118-M10
10578-528EA 335 USD
10578-528
Anti-CAMK1D Mouse Monoclonal Antibody [clone: 3H8]
Antibodies
Type: Primary
Antigen: CAMK1D
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR