Anti-CTNNBIP1 Mouse Monoclonal Antibody [clone: 3B3]

Supplier: Abnova
H00056998-M03
10583-180EA 335 USD
10583-180
Anti-CTNNBIP1 Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Type: Primary
Antigen: CTNNBIP1
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR