Anti-TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast)) Mouse Monoclonal Antibody [clone: 4G4]

Supplier: Abnova
H00056993-M01
10604-392EA 335 USD
10604-392
Anti-TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast)) Mouse Monoclonal Antibody [clone: 4G4]
Antibodies
Type: Primary
Antigen: TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast))
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR