Anti-TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast)) Mouse Polyclonal Antibody

Supplier: Abnova
H00056993-A03
10572-744EA 243 USD
10572-744
Anti-TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast)) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR