Anti-SHISA5 Mouse Monoclonal Antibody [clone: 3G5]

Supplier: Abnova
H00051246-M03
10600-764EA 335 USD
10600-764
Anti-SHISA5 Mouse Monoclonal Antibody [clone: 3G5]
Antibodies
Type: Primary
Antigen: SHISA5
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR