Anti-VRK3 Mouse Polyclonal Antibody

H00051231-B01
10573-812EA 335 USD
10573-812
Anti-VRK3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VRK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR