Anti-DUSP13 Mouse Polyclonal Antibody

H00051207-A01
10555-458EA 243 USD
10555-458
Anti-DUSP13 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DUSP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR