Anti-RSL24D1 Mouse Monoclonal Antibody [clone: 2A10-1E5]

Supplier: Abnova
H00051187-M01
10578-242EA 335 USD
10578-242
Anti-RSL24D1 Mouse Monoclonal Antibody [clone: 2A10-1E5]
Antibodies
Type: Primary
Antigen: RSL24D1
Clonality: Monoclonal
Clone: 2A10-1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR