Anti-DCXR Mouse Polyclonal Antibody

H00051181-A01
10554-788EA 243 USD
10554-788
Anti-DCXR Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DCXR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR