Anti-LEF1 Mouse Monoclonal Antibody [clone: 3A2]

Supplier: Abnova
H00051176-M10
10592-378EA 335 USD
10592-378
Anti-LEF1 Mouse Monoclonal Antibody [clone: 3A2]
Antibodies
Type: Primary
Antigen: LEF1
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR