Anti-HN1 Mouse Monoclonal Antibody [clone: 2C8]

Supplier: Abnova
H00051155-M01
10589-072EA 335 USD
10589-072
Anti-HN1 Mouse Monoclonal Antibody [clone: 2C8]
Antibodies
Type: Primary
Antigen: HN1
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR