Anti-TRIM17 Mouse Monoclonal Antibody [clone: 2D3]

Supplier: Abnova
H00051127-M03
10572-926EA 335 USD
10572-926
Anti-TRIM17 Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
Type: Primary
Antigen: TRIM17
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR