Anti-ZC2HC1A Mouse Polyclonal Antibody

Supplier: Abnova
H00051101-B01P
10552-184EA 335 USD
10552-184
Anti-ZC2HC1A Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZC2HC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR