Anti-GAL Mouse Monoclonal Antibody [clone: 3C1-G5]

Supplier: Abnova
H00051083-M01
10586-986EA 335 USD
10586-986
Anti-GAL Mouse Monoclonal Antibody [clone: 3C1-G5]
Antibodies
Type: Primary
Antigen: GAL (galanin/GMAP prepropeptide)
Clonality: Monoclonal
Clone: 3C1-G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR