Anti-GMNN Mouse Monoclonal Antibody [clone: 1A8]

Supplier: Abnova
H00051053-M01
10587-560EA 335 USD
10587-560
Anti-GMNN Mouse Monoclonal Antibody [clone: 1A8]
Antibodies
Type: Primary
Antigen: GMNN (geminin, DNA replication inhibitor)
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR