Anti-FIS1 Mouse Monoclonal Antibody [clone: 1G9]

Supplier: Abnova
H00051024-M01
10553-356EA 335 USD
10553-356
Anti-FIS1 Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
Type: Primary
Antigen: FIS1 (fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR