Anti-STMN3 Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Abnova
H00050861-M01A
10625-268EA 335 USD
10625-268
Anti-STMN3 Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
Type: Primary
Antigen: STMN3
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR