Anti-DHH Mouse Monoclonal Antibody [clone: 2G5]

Supplier: Abnova
H00050846-M07
10583-848EA 335 USD
10583-848
Anti-DHH Mouse Monoclonal Antibody [clone: 2G5]
Antibodies
Type: Primary
Antigen: DHH
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR