Anti-HDGFRP3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00050810-D01P
10588-548EA 376 USD
10588-548
Anti-HDGFRP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDGFRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR