Anti-DCAF8 Mouse Monoclonal Antibody [clone: 1A12]

Supplier: Abnova
H00050717-M01A
10624-402EA 335 USD
10624-402
Anti-DCAF8 Mouse Monoclonal Antibody [clone: 1A12]
Antibodies
Type: Primary
Antigen: DCAF8
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR