Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 4B2]

Supplier: Abnova
H00050674-M06
10594-756EA 335 USD
10594-756
Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 4B2]
Antibodies
Type: Primary
Antigen: NEUROG3 (neurogenin 3)
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR