Anti-RRM2B Mouse Monoclonal Antibody [clone: 6C1]

Supplier: Abnova
H00050484-M01
10570-410EA 335 USD
10570-410
Anti-RRM2B Mouse Monoclonal Antibody [clone: 6C1]
Antibodies
Type: Primary
Antigen: RRM2B
Clonality: Monoclonal
Clone: 6C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR