Anti-CLDN20 Mouse Monoclonal Antibody [clone: 4E6]

Supplier: Abnova
H00049861-M01
10582-344EA 335 USD
10582-344
Anti-CLDN20 Mouse Monoclonal Antibody [clone: 4E6]
Antibodies
Type: Primary
Antigen: CLDN20
Clonality: Monoclonal
Clone: 4000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR