Anti-KLK14 Mouse Monoclonal Antibody [clone: 2A7]

Supplier: Abnova
H00043847-M05
10591-250EA 335 USD
10591-250
Anti-KLK14 Mouse Monoclonal Antibody [clone: 2A7]
Antibodies
Type: Primary
Antigen: KLK14
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR